Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACCN5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981964
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1964 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1964 Supplier Bioss Supplier No. BS12130R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cation channel that gives rise to very low constitutive currents in the absence of activation. The activated channel exhibits selectivity for sodium, and is inhibited by amiloride.

Specifications

Antigen ACCN5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ASIC5
Gene Accession No. Q9NY37
Gene Alias Accn5; acid sensing (proton gated) ion channel family member 5; acid sensing ion channel family member 5; acid sensing ion channel subunit family member 5; acid-sensing (proton-gated) ion channel family member 5; Acid-sensing ion channel 5; Amiloride-sensitive cation channel 5; amiloride-sensitive cation channel 5, intestinal; amiloride-sensitive Na+ channel; amiloride-sensitive Na+ channel (BLINaC gene); amiloride-sensitive sodium channel; ASIC5; Blinac; Brain-liver-intestine amiloride-sensitive Na(+) channel; brain-liver-intestine amiloride-sensitive sodium channel; HINAC; Human intestine Na(+) channel; INAC; inactivation no afterpotential C
Gene Symbols ASIC5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ACCN5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51802
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.