Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACCS Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553870
Description
Immunogen sequence: NHARLKAAHT YVSEELRALG IPFLSRGAGF FIWVDLRKYL PKGTFEEEML LWRRFLDNKV LLSFGKAFEC KEPG Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
Does not catalyze the synthesis of 1-aminocyclopropane- 1-carboxylate but is capable of catalyzing the deamination of L- vinylglycine.
Specifications
ACCS | |
Polyclonal | |
Unconjugated | |
ACCS | |
1-aminocyclopropane-1-carboxylate synthase (non-functional); 1-aminocyclopropane-1-carboxylate synthase homolog; 1-aminocyclopropane-1-carboxylate synthase homolog (Arabidopsis)(non-functional); 1-aminocyclopropane-1-carboxylate synthase homolog (inactive); 1-aminocyclopropane-1-carboxylate synthase-like protein (non-functional) isoform 1; 1-aminocyclopropane-1-carboxylate synthase-like protein (non-functional) isoform 2; 1-aminocyclopropane-1-carboxylate synthase-like protein (non-functional) isoform 4; 1-aminocyclopropane-1-carboxylate synthase-like protein 1; 2610203E10Rik; ACC synthase-like protein 1; ACCS; ACS; Phacs; RGD1309314 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
311218, 329470, 84680 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A2AIG8, Q96QU6 | |
ACCS | |
Recombinant protein corresponding to Human ACCS. Recombinant protein control fragment (Product #RP-92405). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction