Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACCS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117097
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117097 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117097 Supplier Invitrogen™ Supplier No. PA5117097
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: THP-1, Jurkat, 293T, Mouse testis, Mouse kidney, Rat kidney Immunogen sequence: LDTRPFQLTV EKLEMALREA HSEGVKVKGL ILISPQNPLG DVYSPEELQE YLVFAKRHRL HVIVDEVYML SVFEKSVGYR SVLSLERLPD PQRTHVMWAT SKDFGMSGLR FGTLYTENQD VATAVASLCR YHGLSGLVQY QMAQLLRDRD WINQVYLPEN HARLKAAHTY VSEELRALGI PFLSRGAGFF IWVDLRKYLP KGTFEEEMLL WRRFLDNKVL LSFGKAFECK EPGWFRFVFS DQVHRLCLGM QRVQQVLAGK SQVAEDPRPS QSQEPSDQRR.

Does not catalyze the synthesis of 1-aminocyclopropane- 1-carboxylate but is capable of catalyzing the deamination of L- vinylglycine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACCS
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ACCS
Gene Accession No. A2AIG8, Q96QU6
Gene Alias 1-aminocyclopropane-1-carboxylate synthase (non-functional); 1-aminocyclopropane-1-carboxylate synthase homolog; 1-aminocyclopropane-1-carboxylate synthase homolog (Arabidopsis)(non-functional); 1-aminocyclopropane-1-carboxylate synthase homolog (inactive); 1-aminocyclopropane-1-carboxylate synthase-like protein (non-functional) isoform 1; 1-aminocyclopropane-1-carboxylate synthase-like protein (non-functional) isoform 2; 1-aminocyclopropane-1-carboxylate synthase-like protein (non-functional) isoform 4; 1-aminocyclopropane-1-carboxylate synthase-like protein 1; 2610203E10Rik; ACC synthase-like protein 1; ACCS; ACS; Phacs; RGD1309314
Gene Symbols ACCS
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 222-501 of human ACCS (NP_1159811).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311218, 329470, 84680
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.