Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACCSL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569542 Supplier Invitrogen™ Supplier No. PA569542
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

ACCSL gene ontology annotations related to this gene include biosynthetic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACCSL
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ACCSL
Gene Accession No. Q4AC99
Gene Alias 1-aminocyclopropane-1-carboxylate synthase (inactive)-like; 1-aminocyclopropane-1-carboxylate synthase homolog (Arabidopsis)(non-functional)-like; 1-aminocyclopropane-1-carboxylate synthase homolog (inactive) like; 1-aminocyclopropane-1-carboxylate synthase-like protein 2; ACC synthase-like protein 2; ACCSL; Probable inactive 1-aminocyclopropane-1-carboxylate synthase-like protein 2
Gene Symbols ACCSL
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human ACCSL.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390110
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.