Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Barricade Tapes

Catalog No. 19804925 Shop All AccuformNMC Products
Click to view available options
Color:
Black, Red
Black, Yellow
Red, Black
Displayed Text:
Danger Asbestos Hazard
Danger Keep Out
Police Line Do Not Cross

High-visibility barricade tapes are a quick, temporary and effective way to surround or section-off hazard areas, danger zones and construction sites

  • Size: 7.6cm x 304.8m (3 in. x 1000 ft.)
  • Material: Polyethylene plastic
  • Non-adhesive
  • Self-wound on a roll
  • Unspool, cut easily, and can be fastened with any method
  • Clear, bold warning message that is continuously repeated the length of the roll
  • Material is economical and disposable, yet can be reused

Specifications

Color Black, Yellow
Width (English) 3 in.
Width (Metric) 7.62 cm
Displayed Text Police Line Do Not Cross
Length (English) 1000 ft.
Thickness 3 mil
For Use With (Application) Crowd control
Length (Metric) 304.8 m
Dimensions (L x W) 1000 ft. x 3 in. (304.8 m x 7.62 cm)
Adhesive Type Non-adhesive
Material HDPE
Type Barricade Tape
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.