Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Chemical and Hazardous Safety Labels

Catalog No. 17998264
Click to view available options
Color:
White
Yellow
Displayed Text:
CAUTION Hot
DANGER Flammable
DANGER Hot

Colorful, compact safety signs

  • Size: 8.9 × 12.7cm (3.5 × 5 in.)
  • Material: Adhesive vinyl
  • Language: English

Specifications

Material Vinyl
Quantity 5/Pk.
Height (English) 3.5 in.
Height (Metric) 8.9 cm
Max. Temperature (English) 176°F
Min. Temperature (English) -20°F
Thickness 4 mil
Width (English) 5 in.
Width (Metric) 12.7 cm
Temperature Range (Metric) -29°C to 80°C
Temperature Range (English) -20°F to 176°F
Displayed Text DANGER Hot
Product Type Chemical and hazardous safety label
Color White
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.