Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Chemical Labels

Catalog No. p-3562587 Shop All AccuformNMC Products
Change view
Click to view available options
Displayed Text:
Diesel Fuel
Xylene
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Displayed Text
19-804-566 Diesel Fuel
19-804-567 Xylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-804-566 Supplier Accuform Signs Supplier No. LCHL532
Only null left
Add to Cart
Add to Cart

for effectively communicating hazardous contents by placing proper labeling and Chemical Identification in and around your facility.

Specifications

Certifications/Compliance OSHA 29 CFR 1910.145
Material Vinyl
Quantity 1 Ea.
Mounting Type Self-adhesive
For Use With (Application) Communicating the potential dangers posed by chemicals either in storage or in use
Thickness 6 mil
Temperature Range (Metric) -20°C to +80°C
Temperature Range (English) -20°F to +176°F
Displayed Text Diesel Fuel
Product Type Chemical and Hazardous Safety Label
Color Black, White
Life Span 5 to 7 Years
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.