Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Electrical Safety Labels

Catalog No. p-4247002
Click to view available options
Displayed Text:
DANGER 480 Volts
DANGER Electrical Hazard
DANGER High Voltage

Provide information about potential hazards to keep employees safe when using electrical equipment.

Specifications

Certifications/Compliance OSHA 29 CFR 1910.145
Finish Non-glare
Material Vinyl
Quantity 5/Pk.
For Use With (Application) Provide proper protection in certain areas in order to stay safe on the job or in the for short term outdoor and self sticking applications
Height (English) 3.5 in.
Max. Temperature (English) 176°F
Min. Temperature (English) -20°F
Thickness 4 mil
Width (English) 5 in.
Temperature Range (Metric) -28°C to 80°C
Temperature Range (English) -20°F to 176°F
Displayed Text DANGER High Voltage
Product Type Electrical Hazard Sign
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.