Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs First Aid Labels

Catalog No. 19804583 Shop All AccuformNMC Products
Click to view available options
Dimensions (L x W):
3.5 x 5 in.
5 x 7 in.

This First Aid Kit Inside label includes pictograms to identify location of the kit

Specifications

Certifications/Compliance OSHA 29 CFR 1910.145
Material Vinyl
Quantity 1 Ea.
Size (English) 5 x 7 in.
Size (Metric) 12.7 x 17.78 cm
Mounting Type Self-adhesive
For Use With (Application) Calling attention to the location of first aid equipment that is placed throughout the facility
Dimensions (L x W) 5 x 7 in.
Displayed Text First Aid Kit Inside
Product Type First Aid Safety Label
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.