Accuform Signs Frequent Stops Labels

Manufacturer: ACCUFORM SIGNS LVHR501RFE

 View more versions of this product

Catalog No. 19-821-789

  Check Availability
Add to Cart

    Specifications

    Display Text Frequent Stops
    Format Label
    For Use With (Application) Instruct truck drivers about certain precautions
    Material Reflective Sheeting
    Life Span 5 to 7 Year(s)
    Product Type Traffic Safety Label
    Quantity Each
    Size (English) 8 x 24 in.
    Size (Metric) 20.32 x 60.96cm
    Temperature Range (English) -20 to 176°F
    Temperature Range (Metric) -20 to 80°C
    Thickness (English) 6mil
    Certifications/Compliance OSHA 29 CFR 1910.145

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit