Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Safety Signs

Catalog No. p-4342229 Shop All AccuformNMC Products
Click to view available options
Material:
Aluminum
Polyethylene
Vinyl
Displayed Text:
CAUTION
CAUTION Hot Surface
DANGER Hot Do Not Touch
Product Type:
Safety Sign
Safety Tape

Keep employees safe from harm by effectively communicating hazardous situations to others

Specifications

Material Vinyl
Description Dura Vinyl
Height (English) 3.5 in.
Max. Temperature (English) 176°F
Min. Temperature (English) -20°F
Width (English) 5 in.
Temperature Range (Metric) -29°C to +80°C
Temperature Range (English) -20°F to +176°F
Dimensions (L x W) 3.5 x 5 in.
Displayed Text CAUTION Hot Surface
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.