Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Striped Marking Tapes

Catalog No. p-4518159 Shop All AccuformNMC Products
Click to view available options
Color:
Red
Yellow
Length (English):
54 ft.
108 ft.

Colored tapes are perfect for 5S programs and marking things in the work area.

Specifications

Adhesive Type Permanent Adhesive
Finish Overlaminate clear surface film
Width (English) 3 in.
Certifications/Compliance OSHA 1910.144
Length (English) 108 ft.
For Use With (Equipment) Fire Protection Equipment
Packaging Quantity 1 Roll
Length (Metric) 32.92 m
Thickness 6 mil
Color Yellow
For Use With (Application) Mark specific areas and promote safety by identifying potential hazard areas, marking general and physical hazardous per OSHA 1910.144
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.