Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs U-Channel Posts

Catalog No. p-3563010
Click to view available options
Color:
Green
Silver
Height (English):
72 in.
96 in.
120 in.

Mount signs to these rugged posts

  • Posts can be set in ground
  • Concrete or portable base
  • 9.53mm (0.375 in.) holes on 25.4mm (1 in.) centers along the length

Order Info

3.1m (10 ft.) and larger posts are required to ship via truck. Shipping charges will be applied based upon weight and zone.

Specifications

Certifications/Compliance NCHRP-350
Includes 0.37 in. dia. Hole on 1 in. centers along the length
Quantity 1 Ea.
For Use With (Application) To display signs either in temporary or permanent situations
Height (English) 72 in.
Height (Metric) 1.9 m
Product Type U-channel Sign post
Color Green
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.