Learn More
Invitrogen™ AccuPrime™ SuperMix I
AccuPrime SuperMix I is a ready-to-use mixture of DNA polymerase, accessory proteins, salts, magnesium, and dNTPs for efficient PCR amplification.
$596.00
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
12-342-010
Encompass_Preferred
View Documents
Invitrogen™
12342010
200 Reactions
Each of 1 for $596.00
Only null left
Edge
Add to Cart
Description
Description
AccuPrime SuperMix I is a ready-to-use mixture of DNA polymerase, accessory proteins, salts, magnesium, and dNTPs for efficient PCR amplification. All you have to do is add template and primers, reducing set-up time by half. For accurate PCR performance, AccuPrime SuperMix I sets the standard. You'll amplify your DNA targets cycle after cycle, eliminating troubleshooting and rework.
In addition to a powerful DNA polymerase, AccuPrime SuperMix I includes Platinumanti-DNA polymerase antibodies that inhibit polymerase activity, providing an automatic hot-start, and a thermal stable accessory protein that enhances specific primer-template hybridization during every cycle of PCR. This unique combination allows for success, even under previously suboptimal conditions.
AccuPrime SuperMix I is designed for amplification of genomic DNA amplicons (> 200 bp), plasmid DNA, or cDNA templates. Alternatively, AccuPrime SuperMix II is designed to amplify a broader range of amplicon sizes up to 4 kb.
• Built-in hot start
Order Info
Shipping Condition: Dry Ice
Videos
SDS
Documents
Documents
Product Certifications
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.