Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ ACCURUN™ 305/306 HCV RNA Positive Control

Catalog No. 22554516 Shop All SeraCare Life Sciences Products
Change view
Click to view available options
Type:
ACCURUN 305 Series 100 HCV Positive Control
ACCURUN 305 Series 400 HCV Positive Control
Quantity:
5 x 1 mL
5 x 4 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
22-554-516 ACCURUN 305 Series 400 HCV Positive Control 5 x 4 mL
22-554-138 ACCURUN 305 Series 100 HCV Positive Control 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-554-516 Supplier SeraCare™ Supplier No. 20200085
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enhance quality control and quality assurance

  • Human plasma based controls mimic patient samples
  • Independent controls meet compliance requirements of regulatory agencies and professional organizations
  • Liquid, ready-to-use: no reconstitution or special handling required
  • Extended shelf life means efficiency: allows user to monitor assay performance with same lot over an extended period of time
  • Manufactured from human serum or plasma
  • Reactive for HCV RNA type 1 and nonreactive for HBsAg and antibodies to HIV 1 and HIV 2 and HTLV
  • For in vitro diagnostic use

Specifications

Certifications/Compliance IVD, CE, HC
Content And Storage 2°C to 8°C
Format Vial
Type ACCURUN 305 Series 400 HCV Positive Control
Intended Use Detect and quantitate HCV RNA
Quantity 5 x 4 mL
Source Human Serum or Plasma
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.