Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ ACCURUN™ QC Serology Multi-Marker Positive Controls

Catalog No. p-3126137
Click to view available options
Quantity:
1 x 5 mL
3 x 1 mL
6 x 3.5 mL
Type:
ACCURUN 21 1000 Multi-analyte Positive Control
ACCURUN 21 2000 Multi-analyte Positive Control
ACCURUN 25 ToRCH IgG Positive Control
ACCURUN 30 EBV IgG Positive Control
ACCURUN 31 EBV IgM Positive Control
ACCURUN 40 Multi-analyte Positive Control
ACCURUN 51 Multi-analyte Hepatitis Positive Control 1
ACCURUN 52 Multi-analyte Hepatitis Positive Control
ACCURUN ToRC IgM Positive Control
This item is not returnable. View return policy
This item is not returnable. View return policy

Simplify testing by using one control vial for analytes commonly grouped in one laboratory area

  • Quality control for infectious disease testing laboratories
  • Detect and prevent errors
  • Monitor assay performance
  • Simplify testing procedures
  • Document regulatory compliance
  • Designed to mimic a patient sample
  • Supplied ready to use, requiring no reconstitution or special handling

Specifications

Certifications/Compliance ISO 13485, CE, IVD, Canadian licensed
Content And Storage -20°C
Type ACCURUN 51 Multi-analyte Hepatitis Positive Control 1
Includes HBeAG, HBc IgM, HAV IgM, Stabilizers (EDTA, buffering agents), 0.1% ProClin (5-Chloro-2-Methyl-4-Isothiazolin-3-one and 2-Methyl-4-Isothiazolin-3-one) as preservative
Intended Use Qualitative determination of Hepatitis B e Antigen (HBeAg), IgM Antibodies to Hepatitis B core Antigen (HBc IgM), IgM Antibodies to Hepatitis A Virus (HAV IgM)
Quantity 1 x 5 mL
Source Human Serum or Plasma
Stability 60 Days Open Vial
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.