Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACD Antibody (OTI2B1), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272169FR
Description
ACD Monoclonal specifically detects ACD in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ACD | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
ACD | |
Full length recombinant protein of human ACD (NP_001075955) produced in HEK293T cells. | |
0.1 mL | |
65057.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2B1 | |
Western Blot, Immunohistochemistry | |
adrenocortical dysplasia homolog (mouse), Pip1, PIP1Tint1, POT1 and TIN2 organizing protein, POT1 and TIN2-interacting protein, Ptop, PTOPTpp1, TIN2 interacting protein 1, TINT1adrenocortical dysplasia protein homolog, TPP1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction