Learn More
Invitrogen™ ACD Monoclonal Antibody (OTI2B1)

Description
ACD Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | ACD |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2B1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ACD |
| Gene Accession No. | Q96AP0 |
| Gene Alias | Acd; ACD shelterin complex subunit and telomerase recruitment factor; ACD, shelterin complex subunit and telomerase recruitment factor; adrenocortical dysplasia; adrenocortical dysplasia homolog; adrenocortical dysplasia homolog (mouse); adrenocortical dysplasia protein; Adrenocortical dysplasia protein homolog; PIP1; POT1 and TIN2-interacting protein; PTOP; TIN2 interacting protein 1; TINT1; TPP 1; TPP I; TPP1; TPPI |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.