Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACE/CD143 Antibody (ACE/3763), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308558F
Description
ACE/CD143 Monoclonal antibody specifically detects ACE/CD143 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)Specifications
ACE/CD143 | |
Monoclonal | |
FITC | |
ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA | |
Recombinant fragment (around aa 652-788) of human ACE/CD143 protein (Uniprot: P12821) | |
100 μg | |
Cancer, Cardiovascular Biology, CD Markers, Cell Biology, Endocrinology, Immunology, Signal Transduction, Stem Cells | |
1636 | |
Store at 4°C in the dark. | |
IgG2c κ |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
ACE/3763 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction