Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACE/CD143 Mouse anti-Human, Clone: ACE/3762, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen ACE/CD143
Clone ACE/3762
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122688
SDP
View Documents
Novus Biologicals
NBP30760720UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

ACE/CD143 Monoclonal specifically detects ACE/CD143 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin.
Specifications

Specifications

ACE/CD143
0.2 mg/ml
Western Blot, ELISA, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human
ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA
Recombinant fragment (around aa 652-788) of human ACE/CD143 protein
Primary
Store at 4°C.
ACE/3762
Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
1636
IgG2c κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.