Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACE/CD143 Rabbit anti-Mouse, Rat, Clone: 1C9J3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | ACE/CD143 |
---|---|
Clone | 1C9J3 |
Dilution | Western Blot 1:500 - 1:2000, Immunoprecipitation 1:50 - 1:200 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
ACE/CD143 Monoclonal antibody specifically detects ACE/CD143 in Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
ACE/CD143 | |
Western Blot 1:500 - 1:2000, Immunoprecipitation 1:50 - 1:200 | |
Monoclonal | |
Purified | |
RUO | |
Mouse, Rat | |
ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA | |
A synthetic peptide corresponding to a sequence within amino acids 1200-1306 of human ACE/CD143 (P12821). YFKPLLDWLRTENELHGEKLGWPQYNWTPNSARSEGPLPDSGRVSFLGLDLDAQQARVGQWLLLFLGIALLVATLGLSQRLFSIRHRSLHRHSHGPQFGSEVELRHS | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
1C9J3 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer, Cardiovascular Biology, CD Markers, Cell Biology, Endocrinology, Immunology, Signal Transduction, Stem Cells | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1636 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title