Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578711
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578711 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578711 Supplier Invitrogen™ Supplier No. PA578711
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat lung tissue, mouse lung tissue, human Raji whole cell. IHC: human lung cancer tissue, human thyroid cancer tissue, mouse lung tissue, rat lung tissue IHC-F: human placenta tissue, mouse lung tissue. ICC/IF: A431 cell.

This gene encodes an enzyme involved in catalyzing the conversion of angiotensin I into a physiologically active peptide angiotensin II. Angiotensin II is a potent vasopressor and aldosterone-stimulating peptide that controls blood pressure and fluid-electrolyte balance. This enzyme plays a key role in the renin-angiotensin system. Many studies have associated the presence or absence of a 287 bp Alu repeat element in this gene with the levels of circulating enzyme or cardiovascular pathophysiologies. Two most abundant alternatively spliced variants of this gene encode two isozymes-the somatic form and the testicular form that are equally active. Multiple additional alternatively spliced variants have been identified but their full length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACE
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene Ace
Gene Accession No. P09470, P12821, P47820
Gene Alias ACE; ACE1; ACE-T; angiotensin 1 converting enzyme 1; angiotensin converting enzyme; angiotensin converting enzyme, somatic isoform; angiotensin I converting enzyme; angiotensin I converting enzyme (peptidyl-dipeptidase A) 1; angiotensin I converting enzyme 1; angiotensin I converting enzyme peptidyl-dipeptidase A 1 transcript; angiotensin I-converting enzyme (Dipeptidyl carboxypeptidase 1); angiotensin-converting enzyme; Angiotensin-converting enzyme, soluble form; angiotensin-converting enzyme; LOW QUALITY PROTEIN: angiotensin-converting enzyme; angiotensin-converting enzyme-like; AW208573; carboxycathepsin; CD143; CD143 antigen; DCP; Dcp1; dipeptidyl carboxypeptidase 1; Dipeptidyl carboxypeptidase 1 (Angiotensin I-converting enzyme); dipeptidyl carboxypeptidase I; dipeptidyl peptidase; I79_009670; ICH; kininase II; LOC103158533; MGC26566; MVCD3; peptidase P; peptidyl dipeptidase i; StsRR92; testicular ECA
Gene Symbols Ace
Host Species Rabbit
Immunogen E.coli-derived human ACE recombinant protein (Position: K651-Y864).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11421, 1636, 24310
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.