Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACE2 Recombinant Rabbit Monoclonal Antibody (5X5W10)
GREENER_CHOICE

Catalog No. PIMA536088
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536088 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536088 Supplier Invitrogen™ Supplier No. MA536088
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HepG2, Mouse brain, Rat lung, Rat brain, Rat kidney Immunogen sequence: QSTIEEQAKT FLDKFNHEAE DLFYQSSLAS WNYNTNITEE NVQNMNNAGD KWSAFLKEQS TLAQMYPLQE IQNLTVKLQL QALQQNGSSV LSEDKSKRLN TILNTMSTIY STGKVCNPDN PQECLLLEPG LNEIMANSLD YNERLWAWES WRSEVGKQLR PLYEEYVVLK NEMARANHYE DYGDYWRGDY EVNGVDGYDY SRGQLIEDVE HTFEEIKPLY EHLHAYVRAK LMNAYPSYIS PIGCLPAHLL GDMWGRFWTN LYSLTVPFGQ KPNIDVTDAM VDQAWDAQRI FKEAEKFFVS VGLPNMTQGF WENSMLTDPG NVQKAVCHPT AWDLGKGDFR ILMCTKVTMD DFLTAHHEMG HIQYDMAYAA QPFLLRNGAN EGFHEAVGEI MSLSAATPKH LKSIGLLSPD FQEDNETEIN FLLKQALTIV GTLPFTYMLE KWRWMVFKGE IPKDQWMKKW WEMKREIVGV VEPVPHDETY CDPASLFHVS NDYSFIRYYT RTLYQFQFQE ALCQAAKHEG PLHKCDISNS TEAGQKLFNM LRLGKSEPWT LALENVVGAK NMNVRPLLNY FEPLFTWLKD QNKNSFVGWS TDWSPYADQS IKVRISLKSA LGDKAYEWND NEMYLFRSSV AYAMRQYFLK VKNQMILFGE EDVRVANLKP RISFNFFVTA PKNVSDIIPR TEVEKAIRMS RSRINDAFRL NDNSLEFLGI QPTLGPPNQP PVS.

Angiotensin-converting enzyme 2 (ACE2) plays a central role in vascular, renal, and myocardial physiology. In contrast to its homolog ACE, ACE2 expression is restricted to heart, kidney, and testis. Recently. ACE2 has also been shown to be a functional receptor of the SARS coronavirus. The normal function of ACE2 is to convert the inactive vasoconstrictor angiotensin I (AngI) to Ang1-9 and the active form AngII to Ang1-7, unlike ACE, which converts AngI to AngII. While the role of these vasoactive peptides is not well understood, lack of ACE2 expression in ace2-/ace2- mice leads to severely reduced cardiac contractility, indicating its importance in regulating heart function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACE2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 5X5W10
Concentration 1.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene ACE2
Gene Accession No. Q5EGZ1, Q8R0I0, Q9BYF1
Gene Alias 2010305L05Rik; ACE 2; Ace2; ACEH; ACE-related carboxypeptidase; angiotensin I converting enzyme (peptidyl-dipeptidase A) 2; angiotensin I converting enzyme 2; angiotensin-converting enzyme 2; Angiotensin-converting enzyme homolog; anigotensin I converting enzyme 2; anigotensin-converting enzyme-related carboxypeptidase; metalloprotease MPROT15; peptidyl-dipeptidase A; Processed angiotensin-converting enzyme 2; renal angiotensin-converting enzyme 2; UNQ868/PRO1885
Gene Symbols ACE2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 18-740 of human ACE2 (NP_0685761).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 302668, 59272, 70008
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.