Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACE2 Monoclonal Antibody (OTI2G7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526629
Description
ACE2 Monoclonal Antibody for Western Blot, IHC (P)
Angiotensin-converting enzyme 2 (ACE2) plays a central role in vascular, renal, and myocardial physiology. In contrast to its homolog ACE, ACE2 expression is restricted to heart, kidney, and testis. Recently. ACE2 has also been shown to be a functional receptor of the SARS coronavirus. The normal function of ACE2 is to convert the inactive vasoconstrictor angiotensin I (AngI) to Ang1-9 and the active form AngII to Ang1-7, unlike ACE, which converts AngI to AngII. While the role of these vasoactive peptides is not well understood, lack of ACE2 expression in ace2-/ace2- mice leads to severely reduced cardiac contractility, indicating its importance in regulating heart function.
Specifications
ACE2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BYF1 | |
ACE2 | |
Human recombinant protein fragment corresponding to amino acids 18-237 of ACE2 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2G7 | |
Unconjugated | |
ACE2 | |
2010305L05Rik; ACE 2; Ace2; ACEH; ACE-related carboxypeptidase; angiotensin I converting enzyme (peptidyl-dipeptidase A) 2; angiotensin I converting enzyme 2; angiotensin-converting enzyme 2; Angiotensin-converting enzyme homolog; anigotensin I converting enzyme 2; anigotensin-converting enzyme-related carboxypeptidase; metalloprotease MPROT15; peptidyl-dipeptidase A; Processed angiotensin-converting enzyme 2; renal angiotensin-converting enzyme 2; UNQ868/PRO1885 | |
Mouse | |
Affinity Chromatography | |
RUO | |
59272 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction