Learn More
Invitrogen™ ACE2 Polyclonal Antibody

Description
In Western blot applications, this antibody detects a band at ~90kDa. A suggested positive control is human kidney tissue lysate. PA5-20045 can be used with blocking peptide PEP-0164.
Specifications
Specifications
| Antigen | ACE2 |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ACE2 |
| Gene Accession No. | Q9BYF1 |
| Gene Alias | 2010305L05Rik; ACE 2; Ace2; ACEH; ACE-related carboxypeptidase; angiotensin I converting enzyme (peptidyl-dipeptidase A) 2; angiotensin I converting enzyme 2; angiotensin-converting enzyme 2; Angiotensin-converting enzyme homolog; anigotensin I converting enzyme 2; anigotensin-converting enzyme-related carboxypeptidase; metalloprotease MPROT15; peptidyl-dipeptidase A; Processed angiotensin-converting enzyme 2; renal angiotensin-converting enzyme 2; UNQ868/PRO1885 |
| Gene Symbols | ACE2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.