Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACE2 Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: GEE-1 , Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19519
Description
ACE2 Monoclonal antibody specifically detects ACE2 in Human, Mouse, Rat samples. It is validated for IP, ICC, WB-Ce, IHC-P, IF.Specifications
ACE2 | |
Monoclonal | |
Unconjugated | |
ACE2 | |
Rabbit | |
100 μg | |
59272 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
GEE-1 | |
angiotensin I converting enzyme (peptidyl-dipeptidase A) 2 | |
ACEH, DKFZp434A014 | |
A synthetic peptide corresponding to human ACE2 | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction