Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acepromazine Maleate, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18600165
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail

A1571, 3598-37-6, C19H22N2OS·C4H4O4

Spectrum™ Chemical Acepromazine Maleate, USP is a phenothiazine derivative antipsychotic drug with veterinary sedative and antiemetic properties. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 3598-37-6
Molecular Formula C23H26N2O5S
Molecular Weight (g/mol) 442.53
InChI Key FQRHOOHLUYHMGG-WLHGVMLRSA-N
IUPAC Name (2E)-but-2-enedioic acid; 1-{10-[3-(dimethylamino)propyl]-10H-phenothiazin-2-yl}ethan-1-one
SMILES OC(=O)\C=C\C(O)=O.CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(C=C12)C(C)=O

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Melting Range 136-139°C
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 98 to 101%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.