Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACER3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584834
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584834 Supplier Invitrogen™ Supplier No. PA584834
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DREGYWGPTT STLDWCEENY SVTWYIAEFW NTVSNLIMII PPMFGAVQSV RDGLEKR Highest antigen sequence indentity to the following orthologs: Mouse - 82%, Rat - 82%.

ACER3 is a protein coding gene. Endoplasmic reticulum and Golgi ceramidase that catalyzes the hydrolysis of unsaturated long-chain C18:1-, C20:1- and C20:4-ceramides, dihydroceramides and phytoceramides into sphingoid bases like sphingosine and free fatty acids at alkaline pH. Ceramides, sphingosine, and its phosphorylated form sphingosine-1-phosphate are bioactive lipids that mediate cellular signaling pathways regulating several biological processes including cell proliferation, apoptosis and differentiation. Controls the generation of sphingosine in erythrocytes, and thereby sphingosine-1-phosphate in plasma. Through the regulation of ceramides and sphingosine-1-phosphate homeostasis in the brain may play a role in neurons survival and function. By regulating the levels of proinflammatory ceramides in immune cells and tissues, may modulate the inflammatory response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACER3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACER3
Gene Accession No. Q9NUN7
Gene Alias 1110057L18Rik; 5430429L08Rik; Acer3; alkaline CDase 3; alkaline ceramidase 3; Alkaline dihydroceramidase SB89; alkaline phytoceramidase; AlkCDase 3; aPHC; AV015045; Phca; phytoceramidase, alkaline; RGD1561254
Gene Symbols ACER3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACER3. Recombinant protein control fragment (Product #RP-108561).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55331
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.