Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ACET
GREENER_CHOICESDP

Catalog No. 272810 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-281-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 27-281-0 Supplier Tocris Bioscience™ Supplier No. 2728/10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent kainate antagonist; displays selectivity for GluK1-containing receptors

Potent and selective GluK1 (formerly GluR5) containing kainate receptor antagonist (IC50 = 7 nM) that displays selectivity over GluK2 (formerly GluR6) containing kainate, NMDA, AMPA and group I mGlu receptors. Reversibly blocks induction of NMDA receptor-independent long term potentiation (LTP) in vitro at nanomolar concentrations.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Chemical Identifiers

CAS 936095-50-0
Molecular Formula C20H21N3O6S

Specifications

Quantity 10 mg
Formula Weight 429.45
Purity >98%
Molecular Weight (g/mol) 431.463
Chemical Name or Material ACET
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.