Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetaldehyde, 99%

Manufacturer:  Alfa Aesar 33244AE

Quantity 100mL

 View more versions of this product

Catalog No. AA33244AE


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

75-07-0
44.06
1089
14,39
CH3CHO
MFCD00006991
505984
Acetaldehyde

Specifications

75-07-0
CH3CHO
MFCD00006991
505984
0.780g/mL
-128°C
-27° (-7°F)
98.5%
Liquid
44.06
1089
14,39
20.8°C
100mL
Acetaldehyde

Safety and Handling

SDS & Safety Documents

GHS H Statement
H224-H351-H302-H319-H335

GHS P Statement
P210-P201-P261-P280-P281-P303+P361+P353-P305+P351+P338-P304+P340-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: ACETALDEHYDE

RTECSNumber : AB1925000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit