Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetaldehyde, 99.5%, extra pure

Catalog No. AC149510010
Click to view available options
Quantity:
10 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 75-07-0 | C2H4O | 44.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-07-0
Molecular Formula C2H4O
Molecular Weight (g/mol) 44.04
MDL Number MFCD00006991
InChI Key IKHGUXGNUITLKF-UHFFFAOYSA-N
Synonym ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique
PubChem CID 177
ChEBI CHEBI:15343
IUPAC Name acetaldehyde
SMILES CC=O

Specifications

CAS Min % 99.4
CAS Max % 100.0
Melting Point -123.0°C
Color Colorless
Density 0.7850g/mL
Boiling Point 21.0°C
Flash Point −27°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Odor Pungent
Packaging Glass bottle
Refractive Index 1.3310 to 1.3330 (20°C, 589nm)
Linear Formula CH3CHO
Quantity 1 L
Beilstein 01, 594
Fieser 01,3
Specific Gravity 0.785
Merck Index 15, 40
Solubility Information Solubility in water: > 500g/L (20°C). Other solubilities: soluble in ethanol and acetone
Formula Weight 44.04
Percent Purity 99.5%
Grade Extra Pure
Physical Form Volatile Liquid
Chemical Name or Material Acetaldehyde, Extra Pure
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Extremely flammable liquid and vapor.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
EINECSNumber 200-836-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.