Learn More
Acetaldoxime, syn + anti, 98%
Description
Acetaldoxime is used as an oxygen scavenger in boiler water. It is also used as an intermediate in chemical synthesis and pharmaceuticals. It is involved in the rearrangement reaction to prepare acetamide by using nickel(II) acetate as a catalyst. It acts as a precursor to prepare heterocyclic compound such as spiroisoxazoline. Further, it is used to prepare alkylated(Z)-oximes by deprotonation followed by reaction with benzyl bromide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acetaldoxime is used as an oxygen scavenger in boiler water. It is also used as an intermediate in chemical synthesis and pharmaceuticals. It is involved in the rearrangement reaction to prepare acetamide by using nickel(II) acetate as a catalyst. It acts as a precursor to prepare heterocyclic compound such as spiroisoxazoline. Further, it is used to prepare alkylated(Z)-oximes by deprotonation followed by reaction with benzyl bromide.
Solubility
Soluble in water, ethanol and ether.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Density | 0.969 |
| Boiling Point | 115°C to 117°C |
| Flash Point | 36°C (96°F) |
| Odor | Characteristic |
| Refractive Index | 1.426 |
| Quantity | 50 g |
| UN Number | UN2332 |
| Beilstein | 1209252 |
| Merck Index | 14,42 |
| Solubility Information | Soluble in water,ethanol and ether. |
| Show More |
Safety and Handling
| Hazard Category | H302+H312+H332 |
| Hazard Statement | GHS H Statement H226-H302-H312-H332-H319 Flammable liquid and vapor. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes serious eye irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ACETALDEHYDE OXIME |
| EINECSNumber | 203-479-6 |
| RTECSNumber | AB2975000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.