Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetamide, 99%, for analysis

Catalog No. p-7077379
Click to view available options
Quantity:
250 g
1 kg

CAS: 60-35-5 | C2H5NO | 59.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-35-5
Molecular Formula C2H5NO
Molecular Weight (g/mol) 59.06
InChI Key DLFVBJFMPXGRIB-UHFFFAOYSA-N
Synonym ethanamide, acetic acid amide, methanecarboxamide, acetimidic acid, amide c2, ethanimidic acid, acetamid, amid kyseliny octove, caswell no. 003h, acetimidic acid van
PubChem CID 178
ChEBI CHEBI:49028
IUPAC Name acetamide
SMILES CC(=O)N

Specifications

Melting Point 79.0°C to 81.0°C
Boiling Point 221.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3CONH2
Quantity 250 g
Merck Index 15,44
Solubility Information Solubility in water: 2000g/L (20°C). Other solubilities: 1 g/2 mL alcohol - 1 g/6 mL pyridine, soluble in chloroform, glycerol and hot benzene, slightly soluble in ether
Formula Weight 59.06
Percent Purity 99%
Grade Analysis
Insoluble Matter 0.025% max. (in water)
Chemical Name or Material Acetamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 200-473-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.