Learn More
Acetanilide, 98%
Description
Acetanilide is used as an intermediate in the synthesis of rubber accelerator, dyes and camphor. It is also used in the synthesis of penicillin and other pharmaceutical products. It is involved in the preparation of 4-acetamidobenzenesulfonyl chloride, which is an intermediate during the synthesis of sulfa drugs. Further, it is employed as a experimental photographic developer. In addition to this, it is used to stabilize cellulose ester varnishes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acetanilide is used as an intermediate in the synthesis of rubber accelerator, dyes and camphor. It is also used in the synthesis of penicillin and other pharmaceutical products. It is involved in the preparation of 4-acetamidobenzenesulfonyl chloride, which is an intermediate during the synthesis of sulfa drugs. Further, it is employed as a experimental photographic developer. In addition to this, it is used to stabilize cellulose ester varnishes.
Solubility
Soluble in ethanol, diethyl ether, acetone, benzene. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases.

Specifications
Specifications
Melting Point | 113°C to 116°C |
Density | 1.121 |
Boiling Point | 304°C to 305°C |
Flash Point | 161°C (321°F) |
Assay Percent Range | 98% |
Quantity | 500 g |
Beilstein | 606468 |
Merck Index | 14,50 |
Solubility Information | Soluble in ethanol,diethyl ether,acetone,benzene. Slightly soluble in water. |
Formula Weight | 135.17 |
Show More |
Safety and Handling
Hazard Category | H302 |
Hazard Statement | GHS H Statement H302-H315-H319-H335 |
Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
EINECSNumber | 203-150-7 |
RTECSNumber | AD7350000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.