Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetate Buffer, pH 6.0, EP Buffer Solution 4002200, Ricca Chemical
SDP

Catalog No. 6416 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L

Ep Buffer Solution 4002200

Specifications

Chemical Name or Material Acetate Buffer
Name Note pH 6.0
CAS 7732-18-5, 631-61-8, 64-19-7
CAS Min % 81.0, 16.23, 0.7
CAS Max % 84.0, 16.89, 0.73
Color Colorless
Concentration 1X
For Use With (Application) Product Monographs
pH 6
Physical Form Liquid
Quantity 500 mL
Solution Type Buffer
Packaging Natural Poly Bottle
Grade Laboratory
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.