Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetazolamide 98.0+%, TCI America™
SDP

Catalog No. A2598100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2598100G 100 g
A259825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2598100G Supplier TCI America Supplier No. A2598100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 59-66-5
Molecular Formula C4H6N4O3S2
Molecular Weight (g/mol) 222.24
MDL Number MFCD00003105
InChI Key BZKPWHYZMXOIDC-UHFFFAOYSA-N
Synonym acetazolamide, diamox, acetazolamid, diacarb, glaupax, defiltran, nephramide, acetamox, cidamex, diluran
PubChem CID 1986
ChEBI CHEBI:27690
IUPAC Name N-(5-sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide
SMILES CC(=O)NC1=NN=C(S1)S(N)(=O)=O

Specifications

Melting Point 259°C
Color White
Quantity 100 g
Formula Weight 222.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Acetazolamide
RTECSNumber AC8225000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.