Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, 25% v/v, Reagents

Catalog No. 01336637
Change view
Click to view available options
Quantity:
4 L
15 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-637 15 gal.
01-336-636 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-637 Supplier Reagents Holdings Llc Supplier No. C710060015E
Only null left
Add to Cart
Add to Cart

Acetic Acid, 25% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 64-19-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Ethanoic Acid, Ethylic Acid
IUPAC Name water
SMILES O

Specifications

pH 2.07
Boiling Point 118°C
Color Colorless
Percent Purity 25%
Grade Reagent
Quantity 15 gal.
Packaging Poly Drum
Physical Form Liquid
Specific Gravity 1.0334
Chemical Name or Material Acetic Acid, 25% v/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.