Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        Acetic Acid, 5% v/v (For AOAC Method 953.08), Fisher Chemical™
    
    
    
        
            Shop All Fisherbrand Products
        
    
    
    
    
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                500 mL
                            
                        
                            
                                1 L
                            
                        
                            
                                4 L
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Natural Poly Bottle
                            
                        
                    Specifications
Specifications
| CAS Min % | 94.0, 4.0 | 
| CAS Max % | 96.0, 6.0 | 
| Molecular Weight (g/mol) | Mixture | 
| Percent Purity | 5% v/v | 
| Grade | Laboratory | 
| Quantity | 1 L | 
| Packaging | Natural Poly Bottle | 
| Chemical Name or Material | Acetic Acid, 5% v/v | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction