Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetic Acid, 50% (v/v), Ricca Chemical

Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Specifications
Specifications
CAS Min % | 49.0 |
Color | Colorless |
CAS Max % | 51.0 |
Concentration | 50% (v/v) |
Percent Purity | 50% v/v |
Grade | Laboratory |
Quantity | 4 L |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Chemical Name or Material | Acetic Acid, 50% (v/v) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction