Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, 99.5%, pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
5L
25L
Packaging:
Glass Bottle
Plastic drum
64-19-7
C2H4O2
60.052
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure
176
CHEBI:15366
acetic acid
CC(=O)O
64-19-7
C2H4O2
60.052
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid,ethylic acid,acetic acid, glacial,methanecarboxylic acid,vinegar acid,glacial,acetasol,acide acetique,essigsaeure
176
CHEBI:15366
acetic acid
CC(=O)O
CAS Min % 99.4
CAS Max % 100.0
Assay Percent Range 99.5%
Linear Formula CH3COOH
Beilstein 02, 96
Fieser 02,5; 05,3; 07,1; 08,1; 12,568; 13,45; 15,12
Merck Index 15, 54
Density 1.0480g/mL
Formula Weight 60.05
Physical Form Liquid
Percent Purity 99.5%
Grade Pure
Infrared Spectrum Authentic
Packaging Glass Bottle
Refractive Index 1.3700 to 1.3730
Specific Gravity 1.048
Solubility Solubility in water: miscible. Other solubilities: miscible with alcohol, glycerol, ether and, carbon tetrachloride, practically insoluble in carbon disulfide
Melting Point 16.0°C to 16.5°C
Color Undesignated
Boiling Point 117.0°C to 118.0°C
Quantity 2.5L
Flash Point 40°C
Chemical Name or Material Acetic acid
Show More Show Less

GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger