Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetic Acid-Ammonium Acetate Buffer TS, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
Chemical Name or Material | Acetic Acid-Ammonium Acetate Buffer |
Name Note | Test Solution |
CAS | 7732-18-5, 631-61-8, 64-19-7 |
CAS Min % | 86.0, 6.65, 5.16 |
CAS Max % | 90.0, 6.92, 5.37 |
Color | Colorless |
Concentration | 1X |
For Use With (Application) | Product Monographs |
pH | 4.7 |
Physical Form | Liquid |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction