Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, Environmental Grade, 99% min, Thermo Scientific™

Catalog No. AA38740AP
Click to view available options
:
500mL

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 64-19-7
Molecular Formula C2H4O2
MDL Number MFCD00036152
UN Number UN2789
Merck Index 14,55
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILES CC(=O)O
IUPAC Name acetic acid
Molecular Weight (g/mol) 60.052
PubChem CID 176
ChEBI CHEBI:15366
Formula Weight 60.05
Percent Purity ≤99%
Grade Environmental
Density 1.049g/mL
Quantity 500mL
Flash Point 40° (104°F)
Odor Pungent
Refractive Index 1.3721
Chemical Name or Material Acetic acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H226
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETIC ACID, GLACIAL
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.