Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, glacial, 99.7+% (metals basis)

Catalog No. p-7050932
Click to view available options
Quantity:
100 mL
500 mL

CAS: 64-19-7 | C2H4O2 | 60.05 g/mol

Good polar protic solvent, in its ester form it act as a solvent for inks paints and coatings. Used in laboratory as a solvent for recrystallization, manufacture of camphor, cellulose acetate, polyvinyl acetate. It involves in Wagner-Meerwein rearrangement. Used for preparation of mono-lithium salt of 5-sulfoisophthalic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5
Melting Point 16.6°C
Boiling Point 118.1°C
UN Number UN2789
Merck Index 14,55
Solubility Information Miscible with water,ethanol,acetone,toluene and immiscible in carbon disulphide.
Formula Weight 60.05
Percent Purity ≥99.9%
Density 1.049 g/mL
Quantity 100 mL
Flash Point 40°C (104°F)
Odor Pungent
Physical Form Liquid
Refractive Index 1.3721
Assay (metals basis)
Chemical Name or Material Acetic Acid, Glacial
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H226
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETIC ACID, GLACIAL
EINECSNumber 200-580-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.