Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetic Acid, Glacial ACS AR, Macron Fine Chemicals™
Shop All Avantor Products
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass
SAFEMOR
Specifications
Specifications
| Formula Weight | 60.05 |
| Grade | ACS |
| Quantity | 2.5 L |
| Packaging | SAFEMOR |
| Chemical Name or Material | Acetic Acid, Glacial |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction