Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, Glacial, Aldehyde Free, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02002119
Click to view available options
Quantity:
500 mL
2.5 L
4 L
Packaging:
Clear Glass Bottle
Clear Glass Bottle in Single Shipper
Clear PVC Coated Glass Bottle
Glass Bottle
PVC Coated Glass Bottle
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Formula Weight 60.05
Grade Meets ACS reagent chemical requirements
Quantity 2.5 L
Flash Point 39.00°C
Packaging Glass Bottle
Specific Gravity 1L = 1.05kg
Chemical Name or Material Acetic Acid, Glacial, Aldehyde Free
Recommended Storage IMPORTANT: Material will freeze if stored below 17°C (63°F)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.