Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetic Acid, Glacial (Trace Metal™ Grade), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Specifications
Specifications
Melting Point | 16°C |
Boiling Point | 117°C |
Formula Weight | 60.5 |
Grade | TraceMetal |
Density | 1.048g/cm³ |
Quantity | 2.5 L |
Packaging | Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 1.52 kPa at 20°C |
Chemical Name or Material | Acetic Acid, Glacial |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction