Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, Glacial (TraceMetal™ Grade), Fisher Chemical™
GSA_VA

Catalog No. A5074 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
2.5 L
4 L
  • Ideal for analysis by IC, GF or Flame AA, ICP-OES and ICP-MS
  • Evaluated for 65 metals at < 1 ppb concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in nonbreakable polypropylene bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture available on COA. 45 mm bottle neck size.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5
Melting Point 16°C
Boiling Point 117°C
Color Colorless
Linear Formula CH3COOH
UN Number 2789
Formula Weight 60.05
Grade TraceMetal
Density 1.048g/cm³
Quantity 4 L
Packaging HDPE Bottle
Physical Form Liquid
Vapor Pressure 1.52 kPa at 20°C
Chemical Name or Material Acetic Acid, Glacial
Show More Show Less
DOTInformation DOT Class 8, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.