Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetic Acid, Glacial, U.S.P. - F.C.C., J.T. Baker™
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Glass Bottle, Single ship
pvc coated glass bottle
Specifications
Specifications
Formula Weight | 60.05 |
Grade | FCC/USP |
Quantity | 2.5 L |
Flash Point | 39.00°C |
Packaging | pvc coated glass bottle |
Specific Gravity | 1L = 1.05kg |
Chemical Name or Material | Acetic Acid, Glacial |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction