Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, potassium salt, 97%, pure, anhydrous, ACROS Organics™

Click to view available options
Quantity:
500g
1kg
2.5kg
Packaging:
Plastic bottle
127-08-2
C2H3KO2
98.14
MFCD00012458
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium acetate,acetic acid, potassium salt,diuretic salt,potassium ethanoate,koac,octan draselny czech,potassiumacetate,acetic acid potassium salt,acok,fema no. 2920
517044
CHEBI:32029
potassium;acetate
CC(=O)[O-].[K+]
127-08-2
C2H3KO2
98.14
MFCD00012458
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium acetate,acetic acid, potassium salt,diuretic salt,potassium ethanoate,koac,octan draselny czech,potassiumacetate,acetic acid potassium salt,acok,fema no. 2920
517044
CHEBI:32029
potassium;acetate
CC(=O)[O-].[K+]
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3CO2K
Beilstein 02, 96
Fieser 01,906; 06,475
Merck Index 15, 7726
Solubility Information Solubility in water: 2694g/L (25°C). Other solubilities: soluble in alcohol, insoluble in ether
Formula Weight 98.14
Physical Form Granular Crystalline Powder
Percent Purity 97%
Grade Pure
Infrared Spectrum Authentic
Packaging Plastic bottle
Solubility Solubility in water: 2694g/L (25°C). Other solubilities: soluble in alcohol, insoluble in ether
Color White
Melting Point 292.0°C
Quantity 500g
Flash Point >250°C
Chemical Name or Material Acetic acid, potassium salt
Show More Show Less