Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.8-100.5%, Honeywell Fluka™

Catalog No. 60045759 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 kg
30 kg
4 x 2.5 L
60 kg
6 x 1 L

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, 99.8-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Melting Point 16.2°C (literature)
Boiling Point 118°C
CAS Max % 100.0000%
Assay Percent Range 99.8 to 100.5%
Linear Formula CH3CO2H
UN Number UN2789
Beilstein 506007
Formula Weight 60.05
Grade puriss.
Density 1.049  g/mL at 25 °C (lit.)
Quantity 4 x 2.5 L
Flash Point 40°C
Packaging Poly Bottle
Chemical Name or Material Acetic acid, Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, 99.8-100.5%
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Acetic acid, glacial
EINECSNumber 200-580-7
Recommended Storage Room Temp
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.